Learn More
Greiner Bio-One™ Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry
Specifications
Additive Lithium Heparin
Color Yellow ring, Green cap
Closure Type Safety Pull Cap
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Tube Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Tube Price Quantity & Availability  
22-040-028
View Documents
Greiner Bio-One™
456087
5mL 13 x 100mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
  • Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
  • Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
Specifications
Lithium Heparin
Yellow ring, Green cap
Safety Pull Cap