Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Greiner Bio-One VACUETTE™ QUICKSHIELD Complete PLUS Available on GSA/VA Contract for Federal Government customers only.
Completely assembled unit with QUICKSHIELD disposable safety holder with a “flash” window in the VISIO PLUS needle hub
Manufacturer:  Greiner Bio-One 450235
 View more versions of this product
Catalog No. 22-040-501
Add to cart
Specifications Description & Specifications
Specifications
CE, OSHA
1 in.
21g
Plastic
Yes
Quickshield™ Complete Plus, Visio Plus
25.4mm
4 to 25°C
Safety Tube Holder