Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator Available on GSA/VA Contract for Federal Government customers only.
For routine clinical chemistry, immunohematology, serology or immunology testing
Manufacturer: Greiner Bio-One 456073
 View more versions of this product
Catalog No. 22-040-068
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Material Plastic
    Additive Microscopic silica particles with separation gel
    Color Red cap w/yellow ring
    Capacity (Metric) 13 x 100mm; 5mL
    Closure Type Non-ridged pull cap
  • Description & Specifications
Related Products
Product Content Feedback