Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
For routine clinical chemistry, immunohematology, serology or immunology testing
Manufacturer:  Greiner Bio-One 454228
 View more versions of this product
Catalog No. 22-040-306
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
13 x 75mm; 4mL
Plastic
Gold with gold ring
Non-ridged pull cap
Microscopic silica particles with separation gel