Greiner Bio-One Snap Caps

For recapping 13mm VACUETTE tubes

Manufacturer: Greiner Bio-One 371523

 View more versions of this product

Catalog No. 22-040-304

  Check Availability
Add to Cart

    Specifications

    Color Lavender
    Product Type Snap Cap
    For Use With (Equipment) Recapping 13mm Vacuette™ tubes to reduce the risk of biohazardous contamination
    Size Universal
    Closure Color Lavender
    Material PET

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit