Greiner Bio-One Winged Safety Blood Collection Sets with Holdex™
Pre-attached set with Holdex is a practical, ready-to-use solution
Manufacturer: Greiner Bio-One 450098
 View more versions of this product
Catalog No. 22-040-166
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Blood Collection Set
    Gauge 23
    Length (English) Needle 0.75 in.
    Length (English) Tube 12 in.
    Quantity 24 /pk.
    Includes Holdex Holder
  • Description & Specifications
Related Products
Product Content Feedback