Learn More
Greiner Bio-One™ HOLDEX™ Tube Holder with Luer
Sterile, single-use disposable tube holder with Luer adapter for use in routine specimen withdrawal
Manufacturer:  Greiner Bio-One™ 450263D
Catalog No. 22-405-149
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Off-center plastic cannula at the top of the holder
  • Stainless-steel needle on the underside of the holder
  • Polypropylene holder and adapter
Compliance
Compliant with OSHA guidelines
Specifications
Specifications
CE, OSHA
For use in routine specimen withdrawal from specimen collection bags or bottles through needleless cannula ports
Luer adapter
Polypropylene
Sterile
Yes
Vacuette™ Blood collection needles and Blood collection sets
4 to 25°C
Tube Holder