Greiner Bio-One Blood Collection Tubes

Do not contain additives and can be used for CSF determinations

Manufacturer: Greiner Bio-One 455001

 View more versions of this product

Catalog No. 22-040-284

  Check Availability
Add to Cart

    Specifications

    Product Type Evacuated Blood Tube
    Applications Blood collection
    Additive No Additive
    Color White/Black
    Closure Color Black
    Tube Size 16 × 100mm
    Capacity (English) 0.3 oz.
    Capacity (Metric) 9mL
    Closure Type Non-ridged pull cap

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit