Learn More
Greiner Bio-One Veterinary Needles
Single-sample needle
Manufacturer:  Greiner Bio-One 450069
 View more versions of this product
Catalog No. 22-040-63
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Stainless Steel
Yes
Yes
Quickshield™ safety tube holder
Polypropylene
1.5 in.
Synthetic Rubber
Vacuette™ Single Sample Blood Collection Needle
CE, OSHA
Routine Blood collection for veterinary use only
18g
Pink protective cap
3.81cm
4 to 25°C