Learn More
Eisco™ Greywacke Sedimentary Rock Specimen Products for Science Education
Graywacke is a sedimentary rock composed of large, angular clasts within a fine-grained clay matrix. Formed during underwater avalanches, these rocks are often found on distal ocean shelves.
$1.00 - $10.50
Specifications
Grade K-16
Type Sedimentary Rock
Specimen Size 2-3 cm
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S27587 Products for Science Education
View Documents
Eisco™
ESNG0080
1/Ea. Each for $1.00
Add to cart
S27609 Products for Science Education
View Documents
Eisco™
ESNG0080PK12
12/Pk. Pack of 12 for $10.50
Add to cart
Description
Description
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for identification & classification exercises
  • Useful for advanced study about sedimentary petrology
Specifications
Specifications
K-16
Sedimentary Rock
2-3 cm