Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRF2 Antibody (OTI2F5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245515
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
GRF2 Monoclonal specifically detects GRF2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
GRF2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:400 - 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
C3GGRF2guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene), CRK SH3-binding GNRP, DKFZp781P1719, Guanine nucleotide-releasing factor 2, Protein C3G, Rap guanine nucleotide exchange factor (GEF) 1, rap guanine nucleotide exchange factor 1 | |
Mouse | |
122.6 kDa | |
0.1 mL | |
Signal Transduction | |
2889 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI2F5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RAPGEF1 | |
Full length human recombinant protein of human RAPGEF1(NP_941372) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction