Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRG (Groucho homolog) Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen GRG (Groucho homolog)
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125812
View Documents
Novus Biologicals
NBP31045525UL
25 μg
Each for $106.19
Add to cart
 
NB125811
View Documents
Novus Biologicals
NBP310455100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

GRG (Groucho homolog) Polyclonal antibody specifically detects GRG (Groucho homolog) in Mouse samples. It is validated for Western Blot
Specifications

Specifications

GRG (Groucho homolog)
Western Blot
Purified
RUO
Mouse
AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5
The immunogen is a synthetic peptide directed towards the C-terminal region of MOUSE GRG (Groucho homolog) (NP_034477). Peptide sequence TPLPVGLQPPSLPAVSAGTGLLSLSALGSQTHLSKEDKNGHDGDTHQEDD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Apoptosis, Signal Transduction, Wnt Signaling Pathway
PBS buffer, 2% sucrose
166
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit