Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRGDS Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS2039R
Description
GRGDS forms the cell-binding domain of a glycoprotein, Osteopontin (OPN) . Although the native form of OPN is active in cell attachment assays, it has been observed that thrombin cleavage of OPN causes substantial enhancement of it′s attachment properties. GRGDS synthetic peptide also mimics the cellular binding site of many adhesive proteins in the extracellular matrix and causes rounding and detachment of spread cells. It induces dissociation of alpha-actinin and vinculin from the sites of focal contacts. This cleavage occurs within residues of the GRGDS sequence raising the possibility of thrombin-cleavage further activating OPN by allowing greater accessibility of the GRGDS domain to cell surface receptors.Specifications
GRGDS | |
1 μg/mL | |
Liquid | |
Rabbit | |
IgG | |
Primary | |
100 μL | |
-20°C | |
Antibody |
Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot | |
Unconjugated | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
KLH conjugated GRGDS. | |
Polyclonal | |
Protein A | |
RUO | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title