Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRHPR Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271430AF647

Catalog No. N271430A647


Add to cart

Description

Description

GRHPR Monoclonal antibody specifically detects GRHPR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

GRHPR
OTI2H4
Western Blot, Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
GRHPR
Full length recombinant protein of human GRHPR (NP_036335) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 1.1.1.81, GLXREC 1.1.1.79, glycerate-2-dehydrogenase, GLYD, glyoxylate reductase/hydroxypyruvate reductase, PH2
Mouse
IgG1
0.1 mL
Primary
9380.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit