Learn More
Aviva Systems Biology Corp GRIK2 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP35515_T100
This is a rabbit polyclonal antibody against GRIK2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GRIK2. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55768