Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIK3 Mouse anti-Human, Clone:2B3D1, Abnova™
Mouse monoclonal antibody raised against partial recombinant human GRIK3.
Supplier: Abnova Corporation MAB17935
Description
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics. [provided by RefSeq]Specifications
GRIK3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Immunofluorescence (1:200-1:1000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
GRIK3 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunofluorescence, Western Blot | |
2B3D1 | |
glutamate receptor, ionotropic, kainate 3 | |
In PBS (0.05% sodium azide) | |
EAA5, GLR7, GLUR7, GluR7a | |
Recombinant protein corresponding to amino acids 32-173 of human GRIK3. | |
2899 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction