Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRIK4 Monoclonal Antibody (8H5H9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531746
Description
MA5-31746 has been tested in indirect ELISA.
This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members.
Specifications
GRIK4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q16099 | |
GRIK4 | |
Purified recombinant fragment of human GRIK4 (AA: extra 21-166) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
8H5H9 | |
Unconjugated | |
GRIK4 | |
6330551K01Rik; EAA1; Excitatory amino acid receptor 1; GluK4; GluRgamma1; glutamate ionotropic receptor kainate type subunit 4; glutamate receptor; glutamate receptor ionotropic, kainate 4; glutamate receptor KA1; Glutamate receptor KA-1; glutamate receptor, ionotropic, kainate 4; GRIK; Grik4; KA1; KA-1; kainate receceptor | |
Mouse | |
Protein G | |
RUO | |
2900 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction