Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIN2B Mouse anti-Human, Clone:6E9A8, Abnova™

Mouse monoclonal antibody raised against recombinant human GRIN2B.

Manufacturer:  Abnova Corporation MAB17590

Catalog No. 89-334-014


Add to cart

Description

Description

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq]
Specifications

Specifications

GRIN2B
6E9A8
glutamate receptor, ionotropic, N-methyl D-aspartate 2B
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:100-1:500)
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
GRIN2B
MGC142178, MGC142180, NMDAR2B, NR2B, hNR3
Recombinant protein corresponding to amino acid 27-163 of human GRIN2B from E. coli.
100μg
Antibody
2904
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit