Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIP1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1202842R
Description
GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Hamster samples. It is validated for ImmunofluorescenceSpecifications
GRIP1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Hamster | |
Purified |
Immunofluorescence | |
DyLight 550 | |
glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1. | |
0.1 mL | |
Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
23426 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction