Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Griseofulvin, Micronized, BP, EP, 97-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
126-07-8
C17H19ClO6
354.78
IIUZTXTZRGLYTI-LKUJNTHKSA-N
(2S,2'R)-7-chloro-4,6,6'-trimethoxy-2'-methyl-3H-spiro[1-benzofuran-2,1'-cyclohexane]-3,4'-dione
COC1CC(=O)C[C@@H](C)[C@]11OC2=C(Cl)C(OC)=CC(OC)=C2C1=O
126-07-8
C17H19ClO6
354.78
IIUZTXTZRGLYTI-LKUJNTHKSA-N
(2S,2'R)-7-chloro-4,6,6'-trimethoxy-2'-methyl-3H-spiro[1-benzofuran-2,1'-cyclohexane]-3,4'-dione
COC1CC(=O)C[C@@H](C)[C@]11OC2=C(Cl)C(OC)=CC(OC)=C2C1=O

G1394, 126-07-8, C17H17ClO6

Spectrum™ Griseofulvin, Micronized, BP, EP is an antifungal drug used most often to treat skin infections such as jock itch, athletes foot, ringworm and fungal infections of the scalp, toenails, and fingernails. The BP and EP grades indicate it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical BP and EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. (CAS No. 126-07-8, Molecular Formula: C17H17ClO6) Griseofulvin, Micronized, BP, EP
Packaging Amber Glass Bottle
Sulfated Ash 0.002
Loss on Drying 0.01
Quantity 100 g
Specific Rotation +354° to +364°
CAS Max % 1
Percent Purity 97 to 102%