Learn More
Aviva Systems Biology Corp GRK5 Antibody - middle region (ARP54750_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GRK5. Alternate names: GPRK5ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61300