Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK5 Mouse anti-Human, Mouse, Rat, Clone: S145-20, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527695

Catalog No. PIMA527695


Add to cart

Description

Description

This gene encodes a member of the guanine nucleotide-binding protein-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
Specifications

Specifications

GRK5
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
P34947, Q62833, Q8VEB1
G protein-coupled receptor kinase, Pan-GRK, GRK-pan, GRK
Mouse
IgG1
100 μg
-20°C
Primary
14773, 2869, 59075
Immunocytochemistry, Immunofluorescence, Western Blot
S145-20
Unconjugated
GRK5
Liquid
Grk5
Fusion protein amino acids 381-590 (C-terminus) of mouse GRK5. 91% identical to human, and 99% identical to rat. >55% identity with GRK6 and GRK4. >40% identity with GRK7 and RK.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit