Learn More
GRK5 Mouse anti-Mouse, DyLight 405, Clone: S145-20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242179V
Catalog No. NBP242179V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GRK5
S145-20
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
GRK5
Fusion protein amino acids 381-590 (C-terminus) of mouse GRK5. 91% identical to human, and 99% identical to rat. >55% identity with GRK6 and GRK4. >40% identity with GRK7 and RK.
Protein G purified
Protein Kinase
Primary
2869
Mouse
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.16, FLJ39780, G protein-coupled receptor kinase 5, G protein-coupled receptor kinase GRK5, GPRK5FP2025
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then 55% identity with GRK6 and GRK4. Greater then 40% identity with GRK7 and RK.Detects 70kDa. Cross-reacts with GRK6.