Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRM1 Mouse anti-Human, Clone:5E12D8, Abnova™
Mouse monoclonal antibody raised against partial recombinant human GRM1.
Supplier: Abnova Corporation MAB17943
Description
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. The canonical alpha isoform of the metabotropic glutamate receptor 1 gene is a disulfide-linked homodimer whose activity is mediated by a G-protein-coupled phosphatidylinositol-calcium second messenger system. Alternative splicing results in multiple transcript variants encoding distinct isoforms; some of which may have distinct functions. [provided by RefSeq]Specifications
GRM1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
GRM1 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
5E12D8 | |
glutamate receptor, metabotropic 1 | |
In PBS (0.05% sodium azide) | |
GPRC1A, GRM1A, MGLUR1, MGLUR1A, mGlu1 | |
Recombinant protein corresponding to amino acids 387-486 of human GRM1. | |
2911 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction