Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Grm1, Mouse anti-Rat, ATTO680, Clone: S75-33, Abnova
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18304
Specifications
Grm1 | |
S75-33 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
glutamate receptor, metabotropic 1 | |
Gprc1a | |
Mouse | |
IgG1 | |
RUO | |
Antibody | |
Monoclonal | |
Rat |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
ATTO680 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Liquid | |
Grm1 | |
Recombinant protein corresponding to amino acids 824-1203 at C-terminus of rat Grm5 | |
100 μg | |
Store at -20°C. | |
Primary | |
24414|24418 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title