Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grm1, Mouse anti-Rat, ATTO700, Clone: S75-33, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18305

Catalog No. 89-338-230


Add to cart

Description

Description

Specifications

Specifications

Grm1
S75-33
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
glutamate receptor, metabotropic 1
Gprc1a
Mouse
IgG1
RUO
Antibody
Monoclonal
Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
ATTO700
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Grm1
Recombinant protein corresponding to amino acids 824-1203 at C-terminus of rat Grm5
100 μg
Store at -20°C.
Primary
24414|24418
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit