Learn More
GRO2 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0214-11B27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212227AF594
Catalog No. NP212227A94
Add to cart
Description
Description
GRO2 Monoclonal antibody specifically detects GRO2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
GRO2
MM0214-11B27
Western Blot
chemokine (C-X-C motif) ligand 2, CINC-2a, GRO2 oncogene, GRO2MIP2, GROb, gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP-2a, MIP2AC-X-C motif chemokine 2, SCYB2MIP2-alpha
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CXCL2
Human GRO2
0.1 mL
Primary
2920