Learn More
GRO2 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0209-13N6, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212228AF750
Catalog No. NP212228A70
Add to cart
Description
Description
GRO2 Monoclonal antibody specifically detects GRO2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
GRO2
RM0209-13N6
Western Blot
chemokine (C-X-C motif) ligand 2, CINC-2a, GRO2 oncogene, GRO2MIP2, GROb, gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP-2a, MIP2AC-X-C motif chemokine 2, SCYB2MIP2-alpha
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CXCL2
Mouse GRO2
0.1 mL
Primary
2920