Learn More
Cole-Parmer Grooving Tool for PFA Compression Fittings, 6 mm OD
Manufacturer: Cole-Parmer 3132149
Bolster the sealing capability of PTFE fittings, which withstand pressures to 150 psi when a tool is used.