Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Group III mGlu Receptor Tocriset™

Selection of 5 group III mGlu receptor ligands

Manufacturer:  Tocris Bioscience™ 1828/1 SET

Catalog No. 18-281-SET


Add to cart
This item is not returnable. View return policy

Description

Description

  • Contains the listed products as lyophilised solids or pre-dissolved in DMSO to a stock concentration of 10 mM in 0.5ml
  • Can be used to investigate the pharmacology of Group III mGlu receptors
Specifications

Specifications

1 Set
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit