Learn More
Cell Signaling TechnologySupplier Diversity Partner Human Interleukin-22 (hIL-22) 10 ug DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 8931SC
8931SC , IL-22, a member of the IL-10 family (1,2), is expressed by Th17 CD4+ T cells, activated T cells, Th1 cells and NK cells (3). IL-22 induces proinflammatory responses, drives production of antimicrobial peptides, and is involved in tissue-repair and wound-healing responses (1). The IL-22 receptor is a heterodimer of IL-22R1 and IL-10R2 (4). Expression of IL-22R is restricted to tissue-resident cells, particularly those of epithelial origin, whereas the IL-10R2 chain is expressed in many more cell types. IL-22 induces phosphorylation of Jak1 and Tyk2, leading to activation of Stat3 and, to a lesser extent, Stat1 and Stat5 (1). IL-22 responses can involve activation of the MEK-ERK-RSK, JNK-SAPK, and p38 pathways (1). Elevated levels of IL-22 have been associated with Crohn's disease and rheumatoid arthritis. IL-22 plays an essential role in host response to the pulmonary pathogen Klebsiella pneumoniae (5).
Catalog No. 50-195-068