Learn More
Cell Signaling TechnologySupplier Diversity Partner Human Interleukin-3 (hIL-3) 10 ug DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 8918SC
8918SC , IL-3 is produced by T cells, mast cells and eosinophils (1). Target cells include hematopoietic progenitors, neutrophils, macrophages, mast cells, eosinophils, lymphoid and erythroid cells (1). The IL-3 receptor is a heterodimer of the IL-3 specific α-chain and the common β-chain, βc, which is also used by GM-CSF and IL-5. (1). Binding of IL-3 can also involve substitution of the βc by a βIL-3-chain that appears to be specific for IL-3 (1,2). Binding of IL-3 to its cognate receptor(s) induces activation of Jak2, phosphorylation of multiple Stats (1,3,5,6), and the PI3K/Akt pathway (1). IL-3 may play an important role in the development of airway inflammation associated with asthma (3,4,5).
Catalog No. 50-195-020