Learn More
Cell Signaling TechnologySupplier Diversity Partner Human Epidermal Growth Factor (hEGF) 50 ug DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 8916SF
8916SF , EGF is produced by epithelial cells, fibroblasts and many other cell types (1,2). Low molecular weight soluble EGF is generated through proteolysis of a larger ~130,000 molecular weight transmembrane precursor (1,2). Both soluble and membrane forms of EGF are active (2). EGF induces proliferation, differentiation, and survival of many cell types including tumor-derived cells (1,2, 3).There are multiple members of the EGF family and multiple members of the Erb/Her EGF receptor family. EGF binds to ErbB1/HER1 and induces homodimerization or induces heterodimerization with other Erb/Her members (1). Binding of EGF signals through the MAPK, PI3K/Akt, and STAT 5 pathways (1). EGF, EGF family members, EGF receptors and their signaling pathways are involved in many cancers and are targets for therapeutic intervention (1, 2).
Catalog No. 50-195-013