Learn More
Cell Signaling TechnologySupplier Diversity Partner Human Insulin-like Growth Factor I (hIGF-I) 50 ug DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 8917SF
8917SF , Most circulating endocrine acting IGF-I is produced by hepatocytes, and paracrine or autocrine acting IGF-I is produced by defined cell types within specific tissues (1,2). Many neoplastic cells produce IGF-I, which regulates a number of cellular processes including energy metabolism, proliferation, and cell survival (3,4). IGF-I activity is regulated by one or more of the six extracellular IGF-binding proteins (IGFBPs). IGFBPs bind to IGF-I and most inhibit IGF-I binding to IGF-I receptor (IGFIR) (1,2). Some IGFBPs may increase cell responses to IGF-I. Binding of IGF-I to IGFIR activates the Akt, JNK, and Erk pathways (2). IGF-I and IGFIR are frequently expressed by cancer cells and may contribute to the proliferation and viability of a number of cancer types (1,2).
Catalog No. 50-195-017