Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Growth Hormone, Human/hGH Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

$198.00 - $580.00

Specifications

Antigen Growth Hormone, Human/hGH Ab-3
Clone GH01 / #2 + GH02 / #7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1328P
View Documents
Thermo Scientific™
MS1328P
Purified Ab with BSA and Azide 1mL
Each for $580.00
Add to cart
 
MS1328P0
View Documents
Thermo Scientific™
MS1328P0
Purified Ab with BSA and Azide 100μL
Each for $198.00
Add to cart
 
MS1328P1
View Documents
Thermo Scientific™
MS1328P1
Purified Ab with BSA and Azide 500μL
Each for $437.50
Add to cart
 
MS1328R7
View Documents
Thermo Scientific™
MS1328R7
Ready-to-use for IHC (UVLP) 7mL
Each for $417.00
Add to cart
 
MS1328P1ABX
View Documents
Thermo Scientific™
MS1328P1ABX
Ab without BSA/Azide (Frozen) 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1328PABX
View Documents
Thermo Scientific™
MS1328PABX
Ab without BSA/Azide (Frozen) 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1328R1
View Documents
Thermo Scientific™
MS1328R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Growth Hormone, Human/hGH Ab-3, Mouse Monoclonal Antibody.

hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.

Host Species: Mouse

Clone: GH01/#2 + GH02/#7

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human growth hormone

Molecular Weight: ~22 kDa

Positive Control: Recombinant human growth hormone, Human anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Growth Hormone, Human/hGH Ab-3
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG2a kappa
GH01 / #2 + GH02 / #7
Monoclonal
Murine
Steroids Hormones and Growth Factors
Purified recombinant human growth hormone
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit