Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP75/HSPA9B/Mortalin Mouse anti-Human, Rat, Canine, Monkey, Janelia Fluor™ 646, Clone: OTI9F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270963JF646

Catalog No. N270963J646


Add to cart

Description

Description

GRP75/HSPA9B/Mortalin Monoclonal antibody specifically detects GRP75/HSPA9B/Mortalin in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GRP75/HSPA9B/Mortalin
OTI9F8
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
HSPA9
Full length recombinant protein of human GRP75/HSPA9B/Mortalin (NP_004125) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
3313.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit