Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP75/HSPA9B/Mortalin Mouse anti-Mouse, FITC, Clone: S52A-42, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121670F
Description
GRP75/HSPA9B/Mortalin Monoclonal antibody specifically detects GRP75/HSPA9B/Mortalin in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GRP75/HSPA9B/Mortalin | |
S52A-42 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Purified | |
HSPA9 | |
Fusion protein amino acids 551-766 of mouse SALM2. | |
Protein G purified | |
Core ESC Like Genes, Stem Cell Markers | |
Primary | |
3313 | |
Mouse |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS with 0.05% Sodium Azide | |
75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
Mouse | |
IgG1 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
Detects approx 75kDa. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title