Learn More
Enzo Life SciencesSupplier Diversity Partner IMMUNOSET Grp94 ELISA development set (96 wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI960077
The IMMUNOSET Grp94 ELISA development set contains sufficient reagents for the development of 5 x 96 well ELISA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own ELISA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Alternative name: Endoplasmin, Tra1, Hsp90B1, Gp96. Contents: Capture antibody, Standard, Detection antibody, SA-HRP. Species Reactivity: Dog. Application: ELISA, Colorimetric detection. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 3 hours. Crossreactivity: This assay detects Grp94 in cell lysates and microsomes. Cross reactivity with human Calreticulin, PDI, HSP90α, HSP90β, HSP70, hamster Grp78 and bovine Hsc70 is less than 0.035%. Long Term Storage: +4°C
Catalog No. 50-658-681