Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRPEL1 Mouse anti-Human, Clone: OTI6G6, liquid, TrueMAB™

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA812313

Encompass_Preferred

Catalog No. 50-168-4515


Add to cart

Description

Description

GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
Specifications

Specifications

GRPEL1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9HAV7
GrpE, HMGE, mt-GrpENo.1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80273
Western Blot
OTI6G6
Unconjugated
GRPEL1
Liquid
GRPEL1
Human recombit protein fragment corresponding to amino acids 28-217 of human GRPEL1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit