Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRPEL1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5109789

Catalog No. PIPA5109789


Only null left
Add to Cart

Description

Description

Immunogen sequence: MAAQCVRLAR RSLPALALSL RPSPRLLCTA TKQKNSGQNL EEDMGQSEQK ADPPATEKTL LEEKVKLEEQ LKETVEKYKR ALADTENLRQ RSQKLVEEAK LYGIQAFCKD LLEVADVLEK ATQCVPKEEI KDDNPHLKNL YEGLVMTEVQ IQKVFTKHGL LKLNPVGAKF DPYEHEALFH TPVEGKEPGT VALVSKVGYK LHGRTLRPAL VGVVKEA.

GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRPEL1
Polyclonal
Unconjugated
GRPEL1
AA408748; FLJ25609; GREPEL1; GrpE; GrpE like 1, mitochondrial; grpE protein homolog 1, mitochondrial; GrpE#1; GrpE1; GRPEL1; GrpE-like 1; GrpE-like 1, mitochondrial; GrpE-like protein cochaperone; HMGE; mt-GrpE#1; mt-Grpel1; stress-inducible chaperone mt-GrpE#1
Rabbit
Affinity chromatography
RUO
17713, 79563, 80273
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97576, Q99LP6, Q9HAV7
GRPEL1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-217 of human GRPEL1 (NP_079472.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.