Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553077
Description
Immunogen sequence: HQVIARRQEP ALRGSPGPLT PHPCNELGPP ASPRTPRPVR RGSGRRLPRP DSLIFYRQKR DC Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Specifications
GRRP1 | |
Polyclonal | |
Unconjugated | |
FAM110D | |
FAM110D; family with sequence similarity 110 member D; family with sequence similarity 110, member D; glycine/arginine rich protein 1; glycine/arginine-rich protein 1; GRRP1; Protein FAM110D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79927 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAY7 | |
FAM110D | |
Recombinant protein corresponding to Human GRRP1. Recombinant protein control fragment (Product #RP-90785). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction