Learn More
Abnova™ GRWD1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00083743T01
Catalog No. 89-946-656
Add to cart
Specifications
Specifications
293T
BC002440
pCMV-GRWD1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
GRWD1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 49.4kDa
Transient overexpression cell lysate was tested with anti-GRWD1 antibody (Mfr. No. H00083743-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human