Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRXCR1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548422

Catalog No. PA548422


Add to cart

Description

Description

May play a role in actin filament architecture in developing stereocilia of sensory cells.
Specifications

Specifications

GRXCR1
Polyclonal
PBS with 0.09% sodium azide
Q50H32
Glutaredoxin domain-containing cysteine-rich protein 1, GRXCR1, DFNB25
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
433899
Western Blot
Unconjugated
Grxcr1
Liquid
GRXCR1
KLH conjugated synthetic peptide between 83-112 amino acids from the Central region of human GRXCR1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit