Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRXCR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571118
Description
This target displays homology in the following species: Human: 100%.
This gene encodes a protein containing a glutaredoxin domain, which functions in protein S-glutathionylation. A mutation in this gene was found in a family with autoosomal recessive nonsyndromic sensorineural deafness-101.
Specifications
GRXCR2 | |
Polyclonal | |
Unconjugated | |
GRXCR2 | |
DFNB101; glutaredoxin and cysteine rich domain containing 2; glutaredoxin domain-containing cysteine-rich protein 1-like protein; Glutaredoxin domain-containing cysteine-rich protein 2; glutaredoxin, cysteine rich 2; GRXCR1-like protein; GRXCR2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
643226 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NFK2 | |
GRXCR2 | |
synthetic peptide directed towards the C-terminal region of human GRXCR2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction