Learn More
Tocris Bioscience™ GS 143 Available on GSA/VA Contract for Federal Government customers only.
β-TrCP1 ligase inhibitor
$178.40 - $741.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
563610 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5636/10
Each for $178.40
Add to cart
563650 View Documents Tocris Bioscience™
5636/50
Each for $741.00
Add to cart
Description
Description
β-TrCP1 ligase inhibitor. Inhibits IκBα ubiquitination (IC50 = 5.2 μM) without affecting proteasome activity. Blocks LPS-induced expression of inflammatory cytokines in human myelomonocytic cells. Suppresses antigen-induced NFκB expression, inflammation and mucus production in airways of OVA-sensitized mice.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.