Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSA 10, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
300833-95-8
C26H30N2O5
450.535
MDLUYYGRCGDKGL-UHFFFAOYSA-N
gsa 10,gsa-10,propyl 4-1-hexyl-4-hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxamido benzoate,propyl 4-1-hexyl-4-hydroxy-2-oxo-1,2-dihydroquinolin-3-yl carbonyl amino benzoate,propyl 4-1-hexyl-4-hydroxy-2-oxoquinoline-3-amido benzoate,gsa hplc,propyl 4-1-hexyl-2-hydroxy-4-oxoquinoline-3-carbonyl amino benzoate
54683103
propyl 4-[(1-hexyl-4-hydroxy-2-oxoquinoline-3-carbonyl)amino]benzoate
CCCCCCN1C2=CC=CC=C2C(=C(C1=O)C(=O)NC3=CC=C(C=C3)C(=O)OCCC)O
This item is not returnable. View return policy
300833-95-8
C26H30N2O5
450.535
MDLUYYGRCGDKGL-UHFFFAOYSA-N
gsa 10,gsa-10,propyl 4-1-hexyl-4-hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxamido benzoate,propyl 4-1-hexyl-4-hydroxy-2-oxo-1,2-dihydroquinolin-3-yl carbonyl amino benzoate,propyl 4-1-hexyl-4-hydroxy-2-oxoquinoline-3-amido benzoate,gsa hplc,propyl 4-1-hexyl-2-hydroxy-4-oxoquinoline-3-carbonyl amino benzoate
54683103
propyl 4-[(1-hexyl-4-hydroxy-2-oxoquinoline-3-carbonyl)amino]benzoate
CCCCCCN1C2=CC=CC=C2C(=C(C1=O)C(=O)NC3=CC=C(C=C3)C(=O)OCCC)O

Smo receptor agonist

Smoothened (Smo) receptor agonist (EC50 = 1.2 μM). Does not recognize the classic cyclopamine (Cat. No. 1623) binding site. Does not promote Smo translocation to the primary cilium; is strongly potentiated by forskolin and cholera toxin. Displays anti-adipogenic effects in vitro, mediated by a non-canonical Hedgehog signaling pathway. Promotes differentiation of multipotent mesenchymal progenitor cells into osteoblasts.
Quantity 10mg
Formula Weight 450.53
Percent Purity >98%
Chemical Name or Material GSA 10

Recommended Storage : Store at +4°C