Learn More
GSC Go Science Crazy Boyle's Law Apparatus Products for Science Education
Provides an accurate, quantitative result for pressure/volume or air at constant temperatures.
Manufacturer:  GSC Go Science Crazy 14012
Catalog No. S08000
Add to cart
Description
Description
  • Provides an accurate quantitative result for the Pressure/Volume relationship or air at constant temperatures
  • Graduated 0-300cm cylinder connects to pressure gauge
  • Inside pressure measured by rotating know on piston
  • Gas tap allows cylinder to measure outside atmosphere
  • Outside pressure dial is graduated in (0-3.4)× 105Pa.
Specifications
Specifications
Measuring quantitative results for the pressure/volume relationship or air at constant temperature
Includes graduated dial
Boyle's Law Apparatus
SDS
Documents
Product Certifications
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Not a toy. Product intended for ages 13 and up.