Learn More
GSC Go Science Crazy Basic Electricity Kit Products for Science Education
Provide students with insight into understanding simple electrical circuits using various elements. The GSC International Basic Electricity Kit makes understanding easy. It includes many accessories, instructions and a polypropylene storage case.
Manufacturer:  GSC Go Science Crazy CRBD01
Catalog No. S09595
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 18 Flash lamp bulbs, 9 connectors with lamp holders, 1 rectifier, 2 flexible leads with crocodile clips, 2 mounted bell pushes, 1 length bare copper wire 20 gage, 1 plastic covered flexible wire, 4 pencil leads, 1 piece steel wool, 2 flexible leads with 4mm plugs and crocodile clips, 12 plain connectors, 1 connector with rheostat, 1 resistor, 2 crocodile clips, 2 soft iron nails 50mm, 1 length bare Eureka wire 34 SWG, 1 Circular hardboard disc drilled, 1 copper foil.
Description
Description
  • Consists of a collection of various components that can be used to provide students with clear insight into understanding simple electrical circuits using various circuit elements, and many other related concepts
  • Kit comes in specially made durable polystyrene trays that facilitate convenient storage after use, while also keeping the kit organized
Specifications
Specifications
Electrical circuit kit
20 Bulb Holders, 12 Battery Holders, 4 Variable Resistance Switches, 8 Push Key Switches, 4 Universal Clips, 4 Electrode Holders, 4 Two-Way Switches, 12 Short Lead Red Cables, 12 Short Lead Black Cables, 8 Alligator Clamps, 1 Insulated Wire Spool, 1 Bare Wire Spool, 1 Eureka Wire Spool Bare, 4 Small Diodes, 4 Large Resistors, 40 Mini Light Bulbs, 12 Carbon Electrodes, 8 Soft Iron Electrodes, 1 Copper Foil Sheet, and 1 Steel Wool.