Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSC Monoclonal Antibody (1C2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523070
Description
Peptide Sequence: LGYNNYFYGQ LHVQAAPVGP ACCGAVPPLG AQQCSCVPTP PGYEGPGSVL VSPVPHQMLP YMNVGTLSRT ELQLLNQLHC RRKRRHRTIF TDEQLEALEN LFQETKYPD.
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
Specifications
GSC | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P56915 | |
GSC | |
GSC (NP_776248, 78 a.a. ~ 186 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1C2 | |
Unconjugated | |
GSC | |
GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
Mouse | |
Affinity Chromatography | |
RUO | |
145258 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction