Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Mouse anti-Human, Clone: OTI4F11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524889

Catalog No. PIMA524889


Add to cart

Description

Description

This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
Specifications

Specifications

GSC
Monoclonal
0.19 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P56915
homeobox protein goosecoid
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
145258
Immunocytochemistry, Immunofluorescence
OTI4F11
Unconjugated
GSC
Liquid
GSC
Recombit fragment expressed in E.coli corresponding to amino acids 107-257 of goosecoid.
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit